TNBT diformazan

Leverantör: TCI EUROPE
Varning

Synonyms: Tetranitro Blue Tetrazolium Diformazan, 1,1'-(3,3'-Dimethoxy-4,4'-diphenylene)bis-3,5-di(p-nitrophenyl)diformazan

B0282-1G B0282-100MG
TCIAB0282-1GEA 2200 SEK
TCIAB0282-1G TCIAB0282-100MG
TNBT diformazan
TNBT diformazan
Formel: C₄₀H₃₀N₁₂O₁₀
MW: 838,75 g/mol
MDL Number: MFCD00059831
CAS nummer: 19333-63-2
UN: 1325
ADR: 4.1,III

Order Now

Specifikation testresultat

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 513.0 to 517.0 nm (DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR