3,5-Diphenyl-1-(1-naphthyl)formazan ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
Violet Tetrazolium Formazan, Tetrazolium Violet Formazan
B0281-1G
B0281-100MG
TCIAB0281-1GEA
1275
SEK
TCIAB0281-1G
TCIAB0281-100MG
3,5-Diphenyl-1-(1-naphthyl)formazan ≥97.0%
3,5-Diphenyl-1-(1-naphthyl)formazan
Formel:
C₂₃H₁₈N₄ MW: 350,43 g/mol |
MDL Number:
MFCD00059534 CAS nummer: 1719-72-8 |
Specifikation testresultat
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...