2-Methyltetrahydrothiophene 98,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

B0257-10ML B0257-25ML B0257-5ML
TCIAB0257-10MLEA 3150 SEK
TCIAB0257-10ML TCIAB0257-25ML TCIAB0257-5ML
2-Methyltetrahydrothiophene 98,0 GC_ANALYSMETOD
2-Methyltetrahydrothiophene
Formel: C₅H₁₀S
MW: 102.20 g/mol
Kokpunkt: 132°C
MDL Number: MFCD00015539
CAS nummer: 1795-09-1
EINECS: 217-270-2
UN: 1993
ADR: 3 II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR