ɑ,ε-Dibenzoyl-DL-lysine ≥99.0%
Leverantör: TCI EUROPE
B0208-1G
B0208-5G
TCIAB0208-1GEA
605
SEK
TCIAB0208-1G
TCIAB0208-5G
ɑ,ε-Dibenzoyl-DL-lysine ≥99.0%
alpha,epsilon-Dibenzoyl-DL-lysine
Formel:
C₂₀H₂₂N₂O₄ MW: 354,41 g/mol |
MDL Number:
MFCD00059684 CAS nummer: 32513-92-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Melting point | 145.0 to 147.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...