N-Benzoyl-D-glucosamine ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
N-((2R,3R,4S,5R)-3,4,5,6-Tetrahydroxy-1-oxohexan-2-yl)benzamide, N-Benzoyl-D-glucosamine, 2-Benzamido-2-deoxy-D-glucopyranose
B0200-25G
B0200-5G
TCIAB0200-25GEA
1725
SEK
TCIAB0200-25G
TCIAB0200-5G
N-Benzoyl-D-glucosamine ≥98.0%
N-Benzoyl-D-glucosamine
Formel:
C₁₃H₁₇NO₆ MW: 283,28 g/mol |
MDL Number:
MFCD00070521 CAS nummer: 655-42-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +40.0 to +45.0 deg (C=2, H2O) (after 24h) |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...