3-Buten-2-ylacetate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 3-Acetoxy-1-butene, 1-Methylallyl Acetate, Acetic Acid 3-Buten-2-yl Ester, But-3-en-2-yl acetate
B0128.0005 B0128-25G B0128-5G
TCIAB0128.0005EA 0 SEK
TCIAB0128.0005 TCIAB0128-25G TCIAB0128-5G
3-Buten-2-ylacetate ≥98.0%
3-Buten-2-ylacetate
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 112°C
MDL Number: MFCD00026202
CAS nummer: 6737-11-7
UN: 3272
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %