2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4,4'-((2-(Hydroxymethyl)phenyl)methylene)diphenol, Phenolphthalol

B0100-5G B0100-25G
TCIAB0100-5GEA 1200 SEK
TCIAB0100-5G TCIAB0100-25G
2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol ≥98.0%
2-[Bis(4-hydroxyphenyl)methyl]benzyl alcohol
Formel: C₂₀H₁₈O₃
MW: 306,36 g/mol
MDL Number: MFCD00059144
CAS nummer: 81-92-5
EINECS: 201-386-5
Merck Index: 14,07245

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 202.0 to 205.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR