2,3-Benzofluorene ≥97.0%
Leverantör: TCI
B0059-100MG B0059-1G
TCIAB0059-100MGEA 615 SEK
TCIAB0059-100MG TCIAB0059-1G
2,3-Benzofluorene ≥97.0%
2,3-Benzofluorene
Formel: C₁₇H₁₂
Kokpunkt: 402°C
MDL Number: MFCD00003595
CAS nummer: 243-17-4
EINECS: 205-952-2
Order Now
Specifikation testresultat
Appearance White to Green to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 209.0 to 214.0 deg-C