Sodium benzenesulphonate ≥96.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Benzenesulphonic acid Sodium Salt, Benzenesulphonic acid, sodium salt, Benzenesulphonate sodium

B0035-25G B0035-500G B0035-100G
TCIAB0035-25GEA 270 SEK
TCIAB0035-25G TCIAB0035-500G TCIAB0035-100G
Sodium benzenesulphonate ≥96.0% (by HPLC, titration analysis)
Sodium benzenesulphonate
Formel: C₆H₆O₃S
MW: 180,15 g/mol
MDL Number: MFCD00065179
CAS nummer: 515-42-4
EINECS: 208-198-2
Merck Index: 14,01070

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Ion exchange titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR