NAM [=N-(9-Acridinyl)maleimide] ≥98.0% for HPLC labeling
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
9-Maleimidoacridine, N-(9-Acridinyl)maleimide
A5591-100MG
A5591-50MG
TCIAA5591-100MGEA
3025
SEK
TCIAA5591-100MG
TCIAA5591-50MG
NAM [=N-(9-Acridinyl)maleimide] ≥98.0% for HPLC labeling
NAM [=N-(9-Acridinyl)maleimide]
Formel:
C₁₇H₁₀N₂O₂ MW: 274,28 g/mol |
MDL Number:
MFCD00042983 CAS nummer: 49759-20-8 |
Specifikation testresultat
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Effect test of HPLC derivatization | Effective as labeling agent for N-Acetyl-L-cysteine |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...