Bis(2,4,6-trichlorophenyl)oxalate ≥98.0% chemiluminescence reagent
Leverantör: TCI EUROPE
Synonyms:
Bis(2,4,6-trichlorophenyl) oxalate, TCPO, Oxalic Acid Bis(2,4,6-trichlorophenyl) Ester
A5302-1G
A5302-5G
TCIAA5302-1GEA
415
SEK
TCIAA5302-1G
TCIAA5302-5G
Bis(2,4,6-trichlorophenyl)oxalate ≥98.0% chemiluminescence reagent
Bis(2,4,6-trichlorophenyl)oxalate
Chemiluminescence reagent for the determination of fluorescent compounds
Formel:
C₁₄H₄Cl₆O₄ MW: 448,88 g/mol |
MDL Number:
MFCD00043061 CAS nummer: 1165-91-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 188.0 to 193.0 deg-C |
Solubility in hot Toluene | almost transparency |
Chemiluminescence | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...