AMI-331 hydrochloride ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 3-Bromo-1-(prop-2-yn-1-yl)-2-(pyridin-4-yl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one hydrochloride

A3352-25MG A3352-5MG
TCIAA3352-25MGEA 5725 SEK
TCIAA3352-25MG TCIAA3352-5MG
AMI-331 hydrochloride ≥97.0%
AMI-331 hydrochloride

Order Now

Specifikation testresultat

Appearance White to Amber powder to crystal
Purity (HPLC) min. 97.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR