Acetylthio-PEG3-C2-Azide 98,0 GC_ANALYSMETOD
Leverantör: TCI
Synonyms: Ethanethioic acid S-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl] ester, S-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl] Ethanethioate
A3319-250MG A3319-50MG
TCIAA3319-250MGEA 11075 SEK
TCIAA3319-250MG TCIAA3319-50MG
Acetylthio-PEG3-C2-Azide 98,0 GC_ANALYSMETOD
Acetylthio-PEG3-C2-Azide
Formel: C₁₀H₁₉N₃O₄S
MW: 277.34 g/mol
Storage Temperature: Frys
MDL Number: MFCD22201539
CAS nummer: 1310827-26-9
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %