2-Acetylamino-4-methylthiazole-5-sulphonyl chloride 98,0 HPLC, T

Leverantör: TCI
Fara

Synonyms: 2-Acetylamino-4-methyl-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-1,3-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methylthiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-5-thiazolesulphonyl chloride

A3288-1G A3288-5G
TCIAA3288-1GEA 710 SEK
TCIAA3288-1G TCIAA3288-5G
2-Acetylamino-4-methylthiazole-5-sulphonyl chloride 98,0 HPLC, T
2-Acetylamino-4-methylthiazole-5-sulphonyl chloride
Formel: C₆H₇ClN₂O₃S₂
MW: 254.72 g/mol
MDL Number: MFCD00005320
CAS nummer: 69812-29-9
EINECS: 274-127-7
UN: 3261
ADR: 8 II

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR