7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Aza-2-methoxy-1-cycloheptene, O-Methylcaprolactim, 7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
A3282-25G
A3282-5G
TCIAA3282-25GEA
1425
SEK
TCIAA3282-25G
TCIAA3282-5G
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98.0%
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Formel:
C₇H₁₃NO MW: 127,19 g/mol Kokpunkt: 53°C Flampunkt: 51°C |
MDL Number:
MFCD00006938 CAS nummer: 2525-16-8 EINECS: 219-762-2 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...