3-Aminomethyltetrahydrofuran ≥97.0%

Leverantör: TCI
Varning

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

A3239-1G A3239-5G
TCIAA3239-1GEA 535 SEK
TCIAA3239-1G TCIAA3239-5G
3-Aminomethyltetrahydrofuran ≥97.0%
3-Aminomethyltetrahydrofuran
Formel: C₅H₁₁NO
MW: 101.149 g/mol
Kokpunkt: 67°C
Flampunkt: 58°C
MDL Number: MFCD08234925
CAS nummer: 165253-31-6
UN: 1993
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR