4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥98.0% free radical, purified by sublimation

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine

A3235-200MG A3235-1G
TCIAA3235-200MGEA 485 SEK
TCIAA3235-200MG TCIAA3235-1G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥98.0% free radical, purified by sublimation
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Formel: C₉H₁₉N₂O
MW: 171,26 g/mol
Kokpunkt: 98°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006479
CAS nummer: 14691-88-4
EINECS: 238-738-2

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Average active Oxygen min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR