Sanguinarine chloride hydrate ≥98.0%
Leverantör: TCI
|
Varning
|
A3229-10MG
A3229-50MG
TCIAA3229-10MGEA
2200
SEK
TCIAA3229-10MG
TCIAA3229-50MG
Sanguinarine chloride hydrate ≥98.0%
Sanguinarine chloride hydrate
Formel:
C₂₀H₁₄ClNO₄·xH₂O Storage Temperature: Kylskåp |
CAS nummer:
1099738-80-3 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Water | 5.0 to 15.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...