3-Amino-α-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
m-Amino-α-methylbenzyl alcohol, 1-(3-Aminophenyl)ethanol, 3-Aminophenyl methyl carbinol, 3-(1-Hydroxyethyl)aniline
A3221-1G
A3221-5G
TCIAA3221-1GEA
430
SEK
TCIAA3221-1G
TCIAA3221-5G
3-Amino-α-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)
3-Amino-α-methylbenzyl alcohol
Formel:
C₈H₁₁NO Kokpunkt: 169°C |
MDL Number:
MFCD00007798 CAS nummer: 2454-37-7 EINECS: 219-525-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 67.0 to 71.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...