(S)-(-)-3-Amino-2-piperidone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
A3171-1G
A3171-5G
TCIAA3171-1GEA
2450
SEK
TCIAA3171-1G
TCIAA3171-5G
(S)-(-)-3-Amino-2-piperidone ≥98.0%
(S)-(-)-3-Amino-2-piperidone
Formel:
C₅H₁₀N₂O MW: 114,15 g/mol Smältpunkt: 87°C Storage Temperature: Frys |
MDL Number:
MFCD09258624 CAS nummer: 34294-79-6 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Specific rotation [a]20/D | -12.0 to -16.0 deg (C=0.5, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...