2-Aminothiophene-3-carboxamide ≥98.0%

Leverantör: TCI EUROPE
Varning

A3165-1G A3165-5G
TCIAA3165-1GEA 255 SEK
TCIAA3165-1G TCIAA3165-5G
2-Aminothiophene-3-carboxamide ≥98.0%
2-Aminothiophene-3-carboxamide
Formel: C₅H₆N₂OS
MW: 142,1789 g/mol
MDL Number: MFCD00706455
CAS nummer: 14080-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d407959e5714dc71333e2d8cb9d206ff102d995&vid=7d407959e5714dc71333e2d8cb9d206ff102d995&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR