(2-Acetoxyethoxy)methyl Acetate ≥96.0%

Leverantör: TCI

A3163-1G A3163-5G
TCIAA3163-1GEA 335 SEK
TCIAA3163-1G TCIAA3163-5G
(2-Acetoxyethoxy)methyl Acetate ≥96.0%
(2-Acetoxyethoxy)methyl Acetate
Formel: C₇H₁₂O₅
Kokpunkt: 116°C
MDL Number: MFCD09031327
CAS nummer: 59278-00-1
EINECS: 261-686-7

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Acid Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR