Auraptene ≥98.0%
Leverantör: TCI
A3147-100MG
A3147-25MG
TCIAA3147-100MGEA
2700
SEK
TCIAA3147-100MG
TCIAA3147-25MG
Auraptene ≥98.0%
Auraptene
Formel:
C₁₉H₂₂O₃ Smältpunkt: 66°C Storage Temperature: Frys |
MDL Number:
MFCD00075948 CAS nummer: 495-02-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 75.00 to 78.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...