Arprinocid 95,0 HPLC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: Arpocox, 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine

A3146-250MG A3146-50MG
TCIAA3146-250MGEA 2000 SEK
TCIAA3146-250MG TCIAA3146-50MG
Arprinocid 95,0 HPLC_ANALYSMETOD
Arprinocid
Formel: C₁₂H₉ClFN₅
MW: 277.69 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00867210
CAS nummer: 55779-18-5
EINECS: 259-817-8

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 24.00 to 26.50 %
Melting point 242.0 to 247.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR