Triapine ≥98.0%

Leverantör: TCI
Fara

A3132-25MG A3132-100MG
TCIAA3132-25MGEA 1750 SEK
TCIAA3132-25MG TCIAA3132-100MG
Triapine ≥98.0%
Triapine
Formel: C₇H₉N₅S
Smältpunkt: 234°C
Storage Temperature: Frys
MDL Number: MFCD00924747
CAS nummer: 143621-35-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87d9f09fe0ba29decf429c790fefbbc513862488&vid=87d9f09fe0ba29decf429c790fefbbc513862488&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 34.00 to 37.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR