[3-(6-Aminohexylamino)propyl]trimethoxysilane ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine, N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine

A3128-5ML A3128-25ML
TCIAA3128-5MLEA 1450 SEK
TCIAA3128-5ML TCIAA3128-25ML
[3-(6-Aminohexylamino)propyl]trimethoxysilane ≥97.0% (by GC, titration analysis)
[3-(6-Aminohexylamino)propyl]trimethoxysilane
Formel: C₁₂H₃₀N₂O₃Si
MW: 278,47 g/mol
MDL Number: MFCD00054943
CAS nummer: 51895-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0251fce4fcef630344e8c08df6b00f9de1a0b54d&vid=0251fce4fcef630344e8c08df6b00f9de1a0b54d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR