Anthracene-2,6-diyl bis(trifluoromethanesulphonate) ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,6-Bis(trifluoromethanesulphonyloxy)anthracene

A3127-200MG A3127-1G
TCIAA3127-200MGEA 1800 SEK
TCIAA3127-200MG TCIAA3127-1G
Anthracene-2,6-diyl bis(trifluoromethanesulphonate) ≥98.0%
Anthracene-2,6-diyl bis(trifluoromethanesulphonate)
Formel: C₁₆H₈F₆O₆S₂
MW: 474,34 g/mol
CAS nummer: 594838-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22525ab1cb90b38217a64ecff94c2127d5305234&vid=22525ab1cb90b38217a64ecff94c2127d5305234&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR