Atorvastatin Lactone ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 5-(4-Fluorophenyl)-1-[2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide, Atorvastatin Related Compound H
A3114-100MG A3114-25MG
TCIAA3114-100MGEA 3750 SEK
TCIAA3114-100MG TCIAA3114-25MG
Atorvastatin Lactone ≥95.0%
Atorvastatin Lactone
Formel: C₃₃H₃₃FN₂O₄
MW: 540,64 g/mol
Smältpunkt: 161°C
MDL Number: MFCD00899262
CAS nummer: 125995-03-1
Merck Index: 14,00864
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a344c6cce4146428b7c18c3bfd8e674a34d7e6&vid=62a344c6cce4146428b7c18c3bfd8e674a34d7e6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 4.70 to 5.70 %