5-Amino-2,2-difluoro-1,3-benzodioxole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline

A3107-1G A3107-5G
TCIAA3107-1GEA 655 SEK
TCIAA3107-1G TCIAA3107-5G
5-Amino-2,2-difluoro-1,3-benzodioxole ≥98.0% (by GC, titration analysis)
5-Amino-2,2-difluoro-1,3-benzodioxole
Formel: C₇H₅F₂NO₂
MW: 173,12 g/mol
Kokpunkt: 97°C
MDL Number: MFCD00190144
CAS nummer: 1544-85-0

Order Now

Specifikation testresultat

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR