5-Azoniaspiro[4.4]nonane iodide ≥98.0%

Leverantör: TCI EUROPE
Varning

A3093-1G A3093-5G
TCIAA3093-1GEA 735 SEK
TCIAA3093-1G TCIAA3093-5G
5-Azoniaspiro[4.4]nonane iodide ≥98.0%
5-Azoniaspiro[4.4]nonane iodide
Formel: C₈H₁₆IN
MW: 253,13 g/mol
CAS nummer: 45650-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4083afc4cfb0c9f04237daf94d1ae22b62ee0c&vid=bd4083afc4cfb0c9f04237daf94d1ae22b62ee0c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR