4-Aminocyclohexanone hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

A3070-1G A3070-5G
TCIAA3070-1GEA 1100 SEK
TCIAA3070-1G TCIAA3070-5G
4-Aminocyclohexanone hydrochloride ≥98.0%
4-Aminocyclohexanone hydrochloride
Formel: C₆H₁₁NO·HCl
MW: 149,62 g/mol
MDL Number: MFCD06659765
CAS nummer: 675112-40-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR