2-Acetamido-3-methylpyridine ≥98.0%
Leverantör: TCI
Varning
Synonyms: 2-Acetamido-3-picoline, N-(3-Methylpyridin-2-yl)acetamide
A3065-1G A3065-5G
TCIAA3065-1GEA 990 SEK
TCIAA3065-1G TCIAA3065-5G
2-Acetamido-3-methylpyridine ≥98.0%
2-Acetamido-3-methylpyridine
Formel: C₈H₁₀N₂O
MW: 150.18 g/mol
Kokpunkt: 158°C
MDL Number: MFCD00495347
CAS nummer: 7463-30-1
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C