5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-(4-Pyridyl)-1,3,4-thiadiazol-2-amine, 2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole

A3038-1G A3038-5G
TCIAA3038-1GEA 405 SEK
TCIAA3038-1G TCIAA3038-5G
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine ≥98.0% (by GC, titration analysis)
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine
Formel: C₇H₆N₄S
MW: 178,22 g/mol
MDL Number: MFCD00473692
CAS nummer: 2002-04-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07687858922df4596163257d09f9640ba8ad611&vid=b07687858922df4596163257d09f9640ba8ad611&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR