2-Acetylamino-5-bromo-6-methylpyridine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Acetamido-5-bromo-6-methylpyridine, 6-Acetamido-3-bromo-2-methylpyridine
A3032-1G
A3032-5G
TCIAA3032-1GEA
725
SEK
TCIAA3032-1G
TCIAA3032-5G
2-Acetylamino-5-bromo-6-methylpyridine ≥98.0% (by HPLC, titration analysis)
2-Acetylamino-5-bromo-6-methylpyridine
Formel:
C₈H₉BrN₂O MW: 229,0700 g/mol |
MDL Number:
MFCD03909755 CAS nummer: 142404-84-0 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...