2-Amino-4-(2,4-difluorophenyl)thiazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
A3029-1G
A3029-5G
TCIAA3029-1GEA
845
SEK
TCIAA3029-1G
TCIAA3029-5G
2-Amino-4-(2,4-difluorophenyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(2,4-difluorophenyl)thiazole
Formel:
C₉H₆F₂N₂S MW: 212,22 g/mol |
MDL Number:
MFCD00472891 CAS nummer: 105512-80-9 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...