4-Aminocrotonic acid hydrochloride ≥97.0%

Leverantör: TCI
Varning

Synonyms: trans-4-Amino-2-butenoic acid hydrochloride

A3011-10MG A3011-100MG
TCIAA3011-10MGEA 950 SEK
TCIAA3011-10MG TCIAA3011-100MG
4-Aminocrotonic acid hydrochloride ≥97.0%
4-Aminocrotonic acid hydrochloride
Formel: C₄H₇NO₂·HCl
MW: 137.56 g/mol
CAS nummer: 2126899-84-9

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR