2-Amino-5-phenyl-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Amino-5-phenyl-1,3,4-thiadiazol, 5-Phenyl-1,3,4-thiadiazol-2-amine
A3002-1G
A3002-5G
TCIAA3002-1GEA
775
SEK
TCIAA3002-1G
TCIAA3002-5G
2-Amino-5-phenyl-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-phenyl-1,3,4-thiadiazole
Formel:
C₈H₇N₃S MW: 177,23 g/mol |
MDL Number:
MFCD00205278 CAS nummer: 2002-03-1 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...