(R)-β-Phenylalaninol ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-3-Amino-3-phenylpropan-1-ol, (R)-1-Phenyl-3-propanolamine, (R)-3-Amino-3-phenyl-1-propanol

A2999-200MG A2999-1G
TCIAA2999-200MGEA 485 SEK
TCIAA2999-200MG TCIAA2999-1G
(R)-β-Phenylalaninol ≥97.0%
(R)-β-Phenylalaninol
Formel: C₉H₁₃NO
MW: 151,2056 g/mol
Smältpunkt: 77°C
Storage Temperature: Kylskåp
MDL Number: MFCD01311791
CAS nummer: 170564-98-4
UN: 3259
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 93.0 %
Specific rotation [a]20/D +21.0 to +25.0 deg (C=1, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR