(2-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2-(2-Aminophenyl)acetonitrile, 2-Aminobenzyl cyanide, (2-Aminophenyl)acetonitrile

A2997-200MG A2997-1G
TCIAA2997-200MGEA 485 SEK
TCIAA2997-200MG TCIAA2997-1G
(2-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)
(2-Aminophenyl)acetonitrile
Formel: C₈H₈N₂
MW: 132,17 g/mol
MDL Number: MFCD00091082
CAS nummer: 2973-50-4
EINECS: 221-015-0
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR