5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Aminopentanoic acid Hydroiodide, 5-AVAI, Homopiperidinic acid Hydroiodide
A2984-1G
A2984-5G
TCIAA2984-1GEA
1300
SEK
TCIAA2984-1G
TCIAA2984-5G
5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)
5-Aminovaleric acid Hydroiodide
Formel:
C₅H₁₁NO₂·HI MW: 245,06 g/mol |
CAS nummer:
1705581-28-7 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Water | max. 200 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...