(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
Leverantör: TCI
Varning
Synonyms: Ir(acac)(COD), (Acetylacetonato)(1,5-cyclooctadiene)iridium(I), Acetylacetonato(1,5-cyclooctadiene)iridium(I), 1,5-Cyclooctadiene(acetylacetonato)iridium(I), Ir(acac)(cod)
A2981-200MG A2981-1G
TCIAA2981-200MGEA 910 SEK
TCIAA2981-200MG TCIAA2981-1G
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
(1,5-Cyclooctadiene)iridium(I)-2,4-pentanedionate
Formel: C₁₃H₁₉O₂Ir
MW: 399.51 g/mol
Smältpunkt: 155°C
MDL Number: MFCD08273779
CAS nummer: 12154-84-6
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystaline
Elemental analysis (Carbon) 37.10 to 41.00 %