3-Acetamidocoumarin ≥98.0%

Leverantör: TCI

Synonyms: N-(2-Oxo-2H-chromen-3-yl)acetamide

A2972-200MG A2972-1G
TCIAA2972-200MGEA 295 SEK
TCIAA2972-200MG TCIAA2972-1G
3-Acetamidocoumarin ≥98.0%
3-Acetamidocoumarin
Formel: C₁₁H₉NO₃
MW: 203.20 g/mol
MDL Number: MFCD00075556
CAS nummer: 779-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d2a007c9f413a40c37e42196efafb864387aa5&vid=12d2a007c9f413a40c37e42196efafb864387aa5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR