Alibendol ≥98.0%
Leverantör: TCI
Synonyms:
Cebera, 5-Allyl-2-hydroxy-N-(2-hydroxyethyl)-3-methoxybenzamide, 5-Allyl-N-(2-hydroxyethyl)-3-methoxysalicylamide
A2965-1G
A2965-5G
TCIAA2965-1GEA
310
SEK
TCIAA2965-1G
TCIAA2965-5G
Alibendol ≥98.0%
Alibendol
Formel:
C₁₃H₁₇NO₄ MW: 251.28 g/mol |
MDL Number:
MFCD00865165 CAS nummer: 26750-81-2 EINECS: 247-960-9 Merck Index: 14,00244 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...