3-Amino-o-anisic acid ≥98.0%

Leverantör: TCI
Varning

A2938-1G A2938-5G
TCIAA2938-1GEA 1025 SEK
TCIAA2938-1G TCIAA2938-5G
3-Amino-o-anisic acid ≥98.0%
3-Amino-o-anisic acid
Formel: C₈H₉NO₃
MDL Number: MFCD09701221
CAS nummer: 861306-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5104a69ba1625b7ed1721a4a3cfd57848741023&vid=a5104a69ba1625b7ed1721a4a3cfd57848741023&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR