1-(4-Isocyanatophenyl)ethanone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Isocyanic acid 4-Acetylphenyl ester, 4-Acetylphenylisocyanate, 4-Acetylphenyl Isocyanate

A2933-1G A2933-5G
TCIAA2933-1GEA 445 SEK
TCIAA2933-1G TCIAA2933-5G
1-(4-Isocyanatophenyl)ethanone ≥98.0%
1-(4-Isocyanatophenyl)ethanone
Formel: C₉H₇NO₂
MW: 161,16 g/mol
Kokpunkt: 139°C
Smältpunkt: 37°C
Storage Temperature: Kylskåp
MDL Number: MFCD00013879
CAS nummer: 49647-20-3

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR