1-Acetyl-5-bromoindoline ≥98.0%

Leverantör: TCI

Synonyms: 1-(5-Bromoindolin-1-yl)ethanone

A2924-5G A2924-25G
TCIAA2924-5GEA 815 SEK
TCIAA2924-5G TCIAA2924-25G
1-Acetyl-5-bromoindoline ≥98.0%
1-Acetyl-5-bromoindoline
Formel: C₁₀H₁₀BrNO
Storage Temperature: Frys
MDL Number: MFCD00056017
CAS nummer: 22190-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a192ccc70e59d7cef2e4450a4a7b901f69c4a93d&vid=a192ccc70e59d7cef2e4450a4a7b901f69c4a93d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR