1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-Acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate, 1-Acetyl-5-bromo-4-chloro-3-indolyl acetate, 3-Acetoxy-1-acetyl-5-bromo-4-chloroindole
A2923-1G
A2923-5G
TCIAA2923-1GEA
790
SEK
TCIAA2923-1G
TCIAA2923-5G
1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate ≥98.0%
1-Acetyl-5-bromo-4-chloro-1H-indol-3-ylacetate
Formel:
C₁₂H₉BrClNO₃ MW: 330,562 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00040632 CAS nummer: 3030-06-6 EINECS: 221-200-6 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...