4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 4-Amino-5-chlorocoumaran-7-carboxylic acid

A2918-200MG A2918-1G
TCIAA2918-200MGEA 575 SEK
TCIAA2918-200MG TCIAA2918-1G
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by HPLC, titration analysis)
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid
Formel: C₉H₈ClNO₃
MW: 213,62 g/mol
MDL Number: MFCD12923204
CAS nummer: 123654-26-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR