1,3-Diphenyl-1H-pyrazol-5-amine ≥98.0%

Leverantör: TCI
Varning

Synonyms: 5-Amino-1,3-diphenylpyrazole

A2904-1G A2904-5G
TCIAA2904-1GEA 325 SEK
TCIAA2904-1G TCIAA2904-5G
1,3-Diphenyl-1H-pyrazol-5-amine ≥98.0%
1,3-Diphenyl-1H-pyrazol-5-amine
Formel: C₁₅H₁₃N₃
MW: 235.29 g/mol
MDL Number: MFCD00084878
CAS nummer: 5356-71-8

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR