5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96.0%

Leverantör: TCI
Varning

Synonyms: 5-Amino-4-bromo-3-methyl-1-phenylpyrazole

A2901-1G A2901-5G
TCIAA2901-1GEA 725 SEK
TCIAA2901-1G TCIAA2901-5G
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole ≥96.0%
5-Amino-4-bromo-3-methyl-1-phenyl-1H-pyrazole
Formel: C₁₀H₁₀BrN₃
MW: 252.12 g/mol
MDL Number: MFCD04039926
CAS nummer: 69464-98-8

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR